Anti-OXA1L Mouse polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB21863-100EA 4140 DKK
ABCAAB21863-100
Anti-OXA1L Mouse polyclonal antibody
Type: Primary
Antigen: OXA1L (oxidase (cytochrome c) assembly 1-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR