Anti-PLCB1 Mouse polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB21824-100EA 4140 DKK
ABCAAB21824-100
Anti-PLCB1 Mouse polyclonal antibody
Type: Primary
Antigen: PLCB1 (phospholipase C, beta 1 (phosphoinositide-specific))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR