Anti-NOS2 Mouse polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB21775-100EA 4590 DKK
ABCAAB21775-100
Anti-NOS2 Mouse polyclonal antibody
Type: Primary
Antigen: NOS2 (nitric oxide synthase 2, inducible)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR