Anti-CD3 Rabbit Monoclonal Antibody [clone: SP7]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB21703-7EA 6210 DKK
ABCAAB21703-7
Anti-CD3 Rabbit Monoclonal Antibody [clone: SP7]
Antistoffer
Type: Primary
Antigen: CD3
Clonality: monoclonal
Clone: SP7
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR