Anti-NAP1L4 Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB21631-100EA 0 DKK
ABCAAB21631-100
Anti-NAP1L4 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NAP1L4 (nucleosome assembly protein 1-like 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human