Anti-NANOG Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB21624-100EA 4620.07 DKK
ABCAAB21624-100
Anti-NANOG Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NANOG (Nanog homeobox)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human