Anti-CDK5 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB21249-100EA 5430 DKK
ABCAAB21249-100
Anti-CDK5 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: CDK5 (cyclin-dependent kinase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR