Anti-HPV16 Mouse monoclonal antibody [clone: 716-325]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB21163-250EA 4140 DKK
ABCAAB21163-250
Anti-HPV16 Mouse monoclonal antibody [clone: 716-325]
Type: Primary
Antigen: HPV16
Clonality: monoclonal
Clone: 716-325
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR