Anti-APOC3 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB21032-100EA 4396.51 DKK
ABCAAB21032-100
Anti-APOC3 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: APOC3 (apolipoprotein C-III)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR