Anti-Hemagglutinin peptide Goat polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB20996-100EA 4140 DKK
ABCAAB20996-100
Anti-Hemagglutinin peptide Goat polyclonal antibody
Type: Primary
Antigen: Hemagglutinin peptide
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR