Anti-Glucose Oxidase Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB20957-1EA 4140 DKK
ABCAAB20957-1
Anti-Glucose Oxidase Rabbit polyclonal antibody
Type: Primary
Antigen: Glucose Oxidase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR