Anti-Staphylococcus aureus Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB20920-100EA 5870 DKK
ABCAAB20920-100
Anti-Staphylococcus aureus Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Staphylococcus aureus
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR