Anti-Influenza A Virus M1 Goat Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB20910-100EA 4290.63 DKK
ABCAAB20910-100
Anti-Influenza A Virus M1 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Influenza A Virus M1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu