Anti-Lidocaine Mouse monoclonal antibody [clone: 602]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB20887-100EA 4140 DKK
ABCAAB20887-100
Anti-Lidocaine Mouse monoclonal antibody [clone: 602]
Type: Primary
Antigen: Lidocaine
Clonality: monoclonal
Clone: 602
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR