Anti-Neisseria gonorrhoeae Mouse monoclonal antibody [clone: 9161]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB20845-100EA 4250 DKK
ABCAAB20845-100
Anti-Neisseria gonorrhoeae Mouse monoclonal antibody [clone: 9161]
Type: Primary
Antigen: Neisseria gonorrhoeae
Clonality: monoclonal
Clone: 9161
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR