Anti-Hepatitis B Virus Surface Antigen (Ad/Ay) Mouse Monoclonal Antibody [clone: 86C]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB20758-100EA 5330 DKK
ABCAAB20758-100
Anti-Hepatitis B Virus Surface Antigen (Ad/Ay) Mouse Monoclonal Antibody [clone: 86C]
Antistoffer
Type: Primary
Antigen: Hepatitis B Virus Surface Antigen (Ad/Ay)
Clonality: monoclonal
Clone: 86C
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR