Anti-pan Lamin Mouse Monoclonal Antibody [clone: X67+X167+X233]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB20740-5EA 5500 DKK
ABCAAB20740-5
Anti-pan Lamin Mouse Monoclonal Antibody [clone: X67+X167+X233]
Antistoffer
Type: Primary
Antigen: pan Lamin
Clonality: monoclonal
Clone: X67+X167+X233
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR