Anti-Yersinia pestis v antigen Mouse monoclonal antibody [clone: Va52]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB20738-500EA 4250 DKK
ABCAAB20738-500
Anti-Yersinia pestis v antigen Mouse monoclonal antibody [clone: Va52]
Type: Primary
Antigen: Yersinia pestis v antigen
Clonality: monoclonal
Clone: Va52
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR