Anti-HNRNPU Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB20666-100EA 6010 DKK
ABCAAB20666-100
Anti-HNRNPU Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HNRNPU (heterogeneous nuclear ribonucleoprotein U (scaffold attachment factor A))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR