Anti-HIRA Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB20655-100EA 3725.86 DKK
ABCAAB20655-100
Anti-HIRA Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HIRA (histone cell cycle regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse