Anti-Penicillinase Rabbit polyclonal antibody HRP (Horseradish Peroxidase)

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB20545-1EA 4250 DKK
ABCAAB20545-1
Anti-Penicillinase Rabbit polyclonal antibody HRP (Horseradish Peroxidase)
Type: Primary
Antigen: Penicillinase
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR