Anti-Parainfluenza Virus type 1 Mouse monoclonal antibody [clone: BID02]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB20502-100EA 4300 DKK
ABCAAB20502-100
Anti-Parainfluenza Virus type 1 Mouse monoclonal antibody [clone: BID02]
Type: Primary
Antigen: Parainfluenza Virus type 1
Clonality: monoclonal
Clone: BID02
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR