Anti-Hepatitis B Virus Surface Antigen (Ad/Ay) Mouse monoclonal antibody [clone: BID40]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB20463-500EA 4300 DKK
ABCAAB20463-500
Anti-Hepatitis B Virus Surface Antigen (Ad/Ay) Mouse monoclonal antibody [clone: BID40]
Type: Primary
Antigen: Hepatitis B Virus Surface Antigen (Ad/Ay)
Clonality: monoclonal
Clone: BID40
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR