Anti-KIDINS220 Mouse Monoclonal Antibody [clone: P220 1F8/3]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB20211-50EA 5330 DKK
ABCAAB20211-50
Anti-KIDINS220 Mouse Monoclonal Antibody [clone: P220 1F8/3]
Antistoffer
Type: Primary
Antigen: KIDINS220 (kinase D-interacting substrate, 220kDa)
Clonality: monoclonal
Clone: P220 1F8/3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR