Anti-Hairy Mouse Monoclonal Antibody [clone: 1/24]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB20165-50EA 3800.38 DKK
ABCAAB20165-50
Anti-Hairy Mouse Monoclonal Antibody [clone: 1/24]
Antistoffer
Type: Primary
Antigen: Hairy
Clonality: monoclonal
Clone: 42028
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu