Anti-RNF5 Mouse Monoclonal Antibody [clone: LHM 2]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB20156-50EA 5800 DKK
ABCAAB20156-50
Anti-RNF5 Mouse Monoclonal Antibody [clone: LHM 2]
Antistoffer
Type: Primary
Antigen: RNF5 (ring finger protein 5, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone: LHM 2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR