Anti-LEP Mouse monoclonal antibody [clone: BDI170]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB20101-50EA 4360 DKK
ABCAAB20101-50
Anti-LEP Mouse monoclonal antibody [clone: BDI170]
Type: Primary
Antigen: LEP (leptin)
Clonality: monoclonal
Clone: BDI170
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR