Anti-Chlamydia trachomatis Mouse monoclonal antibody [clone: 130]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB20037-200EA 4140 DKK
ABCAAB20037-200
Anti-Chlamydia trachomatis Mouse monoclonal antibody [clone: 130]
Type: Primary
Antigen: Chlamydia trachomatis
Clonality: monoclonal
Clone: 130
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR