Anti-SKI Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB19864-100EA 5670 DKK
ABCAAB19864-100
Anti-SKI Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SKI (v-ski avian sarcoma viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR