Anti-E tag Goat Polyclonal Antibody (FITC (Fluorescein))

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB19399-100EA 5630 DKK
ABCAAB19399-100
Anti-E tag Goat Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Type: Primary
Antigen: E tag
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR