Anti-ANTXR1 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB19387-100EA 5500 DKK
ABCAAB19387-100
Anti-ANTXR1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ANTXR1 (anthrax toxin receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR