Anti-DPYSL5 Rat monoclonal antibody [clone: CR-1]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB19378-100EA 4360 DKK
ABCAAB19378-100
Anti-DPYSL5 Rat monoclonal antibody [clone: CR-1]
Type: Primary
Antigen: DPYSL5 (dihydropyrimidinase-like 5)
Clonality: monoclonal
Clone: CR-1
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR