Anti-HCN2 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB19346-100EA 5760 DKK
ABCAAB19346-100
Anti-HCN2 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: HCN2 (hyperpolarization activated cyclic nucleotide-gated potassium channel 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR