Anti-ESRRB Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB19331-100EA 4290.63 DKK
ABCAAB19331-100
Anti-ESRRB Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ESRRB (estrogen-related receptor beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat