Anti-Mouse Serum Albumin Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB19195-1EA 4098.45 DKK
ABCAAB19195-1
Anti-Mouse Serum Albumin Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Type: Primary
Antigen: Mouse Serum Albumin
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse