Anti-HB Sheep polyclonal antibody AP (Alkaline Phosphatase)

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB19190-100EA 4650 DKK
ABCAAB19190-100
Anti-HB Sheep polyclonal antibody AP (Alkaline Phosphatase)
Type: Primary
Antigen: HB (hemoglobin)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR