Anti-TF Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB19178-1EA 3949.41 DKK
ABCAAB19178-1
Anti-TF Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Type: Primary
Antigen: TF (transferrin)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human