Anti-ZMYM3 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB19165-100EA 5670 DKK
ABCAAB19165-100
Anti-ZMYM3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ZMYM3 (zinc finger, MYM-type 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR