Anti-SERPINC1 Chicken Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB19105-50EA 5430 DKK
ABCAAB19105-50
Anti-SERPINC1 Chicken Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SERPINC1 (serpin peptidase inhibitor, clade C (antithrombin), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR