Anti-Fibrin alpha chain Mouse Monoclonal Antibody [clone: UC45]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB19079-50EA 4290.63 DKK
ABCAAB19079-50
Anti-Fibrin alpha chain Mouse Monoclonal Antibody [clone: UC45]
Antistoffer
Type: Primary
Antigen: Fibrin alpha chain
Clonality: monoclonal
Clone: UC45
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR