Anti-FOLH1 Mouse monoclonal antibody unconjugated [clone: YPSMA-1]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB19071-250EA 5570 DKK
ABCAAB19071-250
Anti-FOLH1 Mouse monoclonal antibody unconjugated [clone: YPSMA-1]
Antistoffer
Type: Primary
Antigen: FOLH1 (folate hydrolase (prostate-specific membrane antigen) 1)
Clonality: monoclonal
Clone: YPSMA-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR