Anti-LAT2 Goat polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB19042-100EA 2710 DKK
ABCAAB19042-100
Anti-LAT2 Goat polyclonal antibody
Type: Primary
Antigen: LAT2 (linker for activation of T cells family, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR