Anti-HSP90AA1 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB19021-100EA 5370 DKK
ABCAAB19021-100
Anti-HSP90AA1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HSP90AA1 (heat shock protein 90kDa alpha (cytosolic), class A member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR