Anti-CXCR5 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB19017-100EA 5370 DKK
ABCAAB19017-100
Anti-CXCR5 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CXCR5 (chemokine (C-X-C motif) receptor 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR