Anti-CCNA2 Mouse monoclonal antibody unconjugated [clone: E43.2]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB18718-50EA 5330 DKK
ABCAAB18718-50
Anti-CCNA2 Mouse monoclonal antibody unconjugated [clone: E43.2]
Antistoffer
Type: Primary
Antigen: CCNA2 (cyclin A2)
Clonality: monoclonal
Clone: E43.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR