Anti-Hepatitis B Virus Core Antigen Mouse monoclonal antibody [clone: 10-E-11]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB18686-100EA 0 DKK
ABCAAB18686-100
Anti-Hepatitis B Virus Core Antigen Mouse monoclonal antibody [clone: 10-E-11]
Type: Primary
Antigen: Hepatitis B Virus Core Antigen
Clonality: monoclonal
Clone: 10-E-11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR