Anti-Hepatitis B Virus Core Antigen Mouse monoclonal antibody unconjugated [clone: C 1]
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB18683-100EA 5140 DKK
ABCAAB18683-100
Anti-Hepatitis B Virus Core Antigen Mouse monoclonal antibody unconjugated [clone: C 1]
Type: Primary
Antigen: Hepatitis B Virus Core Antigen
Clonality: monoclonal
Clone: C 1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestil nu