Anti-Hepatitis C Virus NS3 Mouse monoclonal antibody [clone: 242]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB18669-250EA 4300 DKK
ABCAAB18669-250
Anti-Hepatitis C Virus NS3 Mouse monoclonal antibody [clone: 242]
Type: Primary
Antigen: Hepatitis C Virus NS3
Clonality: monoclonal
Clone: 242
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR