Anti-B2M Mouse Monoclonal Antibody [clone: BBM.1]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB18605-50EA 5670 DKK
ABCAAB18605-50
Anti-B2M Mouse Monoclonal Antibody [clone: BBM.1]
Antistoffer
Type: Primary
Antigen: B2M (beta-2-microglobulin)
Clonality: monoclonal
Clone: BBM.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR