Anti-IL22 Receptor Alpha Goat Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB18568-100EA 5670 DKK
ABCAAB18568-100
Anti-IL22 Receptor Alpha Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: IL22 Receptor Alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR